Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49483629-49483862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf32
Entrez Gene ID
497661
Gene Name
chromosome 18 open reading frame 32
Gene Aliases
-
UniGene
Hs.603040
Gene Chromosome Location
Chr.18: 49481178-49487274 on Build GRCh38
Transcripts
NM_001035005.3 NM_001199346.1 NM_001199355.1 NM_001199356.1

Gene Symbol
RPL17-C18orf32
Entrez Gene ID
100526842
Gene Name
RPL17-C18orf32 readthrough
Gene Aliases
-
UniGene
Hs.374588
Gene Chromosome Location
Chr.18: 49481178-49492565 on Build GRCh38
Transcripts
NM_001035005.3 NM_001199346.1 NM_001199355.1 NM_001199356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4140095 c.31G>C p.V11L Chr.18: 49483718-49483718 on Build GRCh38
6769178 c.93C>T p.F31F Chr.18: 49483656-49483656 on Build GRCh38
709430 c.66A>G p.P22P Chr.18: 49483683-49483683 on Build GRCh38
6148545 c.89C>T p.P30L Chr.18: 49483660-49483660 on Build GRCh38
2878825 c.83T>C p.V28A Chr.18: 49483666-49483666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777139159 Chr.18: 49483739-49483739 on Build GRCh38
rs202115703 Chr.18: 49483710-49483710 on Build GRCh38
rs749060876 Chr.18: 49483674-49483675 on Build GRCh38
rs751544895 Chr.18: 49483680-49483680 on Build GRCh38
rs780243305 Chr.18: 49483699-49483699 on Build GRCh38
rs959774832 Chr.18: 49483820-49483820 on Build GRCh38
rs766028505 Chr.18: 49483655-49483655 on Build GRCh38
rs769307126 Chr.18: 49483757-49483757 on Build GRCh38
rs1041952354 Chr.18: 49483788-49483788 on Build GRCh38
rs377011626 Chr.18: 49483752-49483752 on Build GRCh38
rs774331787 Chr.18: 49483748-49483748 on Build GRCh38
rs143746104 Chr.18: 49483722-49483722 on Build GRCh38
rs767363331 Chr.18: 49483796-49483796 on Build GRCh38
rs748334926 Chr.18: 49483715-49483715 on Build GRCh38
rs867268383 Chr.18: 49483693-49483694 on Build GRCh38
rs747588792 Chr.18: 49483727-49483727 on Build GRCh38
rs752328241 Chr.18: 49483665-49483665 on Build GRCh38
rs775042329 Chr.18: 49483679-49483679 on Build GRCh38
rs758219583 Chr.18: 49483672-49483672 on Build GRCh38
rs375259579 Chr.18: 49483825-49483825 on Build GRCh38
rs762567484 Chr.18: 49483768-49483768 on Build GRCh38
rs759291520 Chr.18: 49483659-49483659 on Build GRCh38
rs561008485 Chr.18: 49483756-49483756 on Build GRCh38
rs761794204 Chr.18: 49483791-49483791 on Build GRCh38
rs751348939 Chr.18: 49483779-49483779 on Build GRCh38
rs764810508 Chr.18: 49483661-49483661 on Build GRCh38
rs748046543 Chr.18: 49483790-49483790 on Build GRCh38
rs375834505 Chr.18: 49483704-49483704 on Build GRCh38
rs1035210902 Chr.18: 49483794-49483794 on Build GRCh38
rs373778829 Chr.18: 49483724-49483724 on Build GRCh38
rs972646924 Chr.18: 49483667-49483667 on Build GRCh38
rs369428991 Chr.18: 49483764-49483764 on Build GRCh38
rs80201970 Chr.18: 49483728-49483728 on Build GRCh38
rs918546080 Chr.18: 49483682-49483682 on Build GRCh38
rs768637811 Chr.18: 49483715-49483718 on Build GRCh38
rs779031886 Chr.18: 49483807-49483807 on Build GRCh38
rs755040185 Chr.18: 49483799-49483799 on Build GRCh38
rs757152863 Chr.18: 49483685-49483685 on Build GRCh38
rs369030829 Chr.18: 49483706-49483706 on Build GRCh38
rs572227147 Chr.18: 49483656-49483656 on Build GRCh38
rs991181511 Chr.18: 49483758-49483758 on Build GRCh38
rs200854465 Chr.18: 49483774-49483774 on Build GRCh38
rs750338395 Chr.18: 49483798-49483798 on Build GRCh38

View Full Product Details