Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88065078-88065351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BANP
Entrez Gene ID
54971
Gene Name
BTG3 associated nuclear protein
Gene Aliases
BEND1, SMAR1, SMARBP1
UniGene
Hs.461705
Gene Chromosome Location
Chr.16: 87949216-88077318 on Build GRCh38
Transcripts
NM_001173539.1 NM_001173540.1 NM_001173541.1 NM_001173542.1 NM_001173543.1 NM_017869.3 NM_079837.2 XM_005256008.3 XM_005256009.3 XM_011523170.2 XM_011523172.2 XM_011523174.2 XM_011523175.2 XM_011523176.2 XM_011523177.2 XM_011523178.2 XM_011523179.2 XM_011523180.2 XM_011523181.2 XM_011523183.2 XM_011523184.2 XM_011523186.2 XM_017023378.1 XM_017023379.1 XM_017023380.1 XM_017023381.1 XM_017023382.1 XM_017023383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1302334 c.1344+1G>T p.? Chr.16: 88065333-88065333 on Build GRCh38
435895 c.1326C>T p.F442F Chr.16: 88065314-88065314 on Build GRCh38
4167365 c.1306C>T p.L436F Chr.16: 88065294-88065294 on Build GRCh38
5385524 c.1254C>T p.S418S Chr.16: 88065308-88065308 on Build GRCh38
6555982 c.1235C>T p.P412L Chr.16: 88065289-88065289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768463634 Chr.16: 88065249-88065249 on Build GRCh38
rs990627332 Chr.16: 88065151-88065151 on Build GRCh38
rs572352552 Chr.16: 88065118-88065118 on Build GRCh38
rs749026758 Chr.16: 88065247-88065247 on Build GRCh38
rs1035182235 Chr.16: 88065137-88065137 on Build GRCh38
rs377676627 Chr.16: 88065312-88065312 on Build GRCh38
rs764593453 Chr.16: 88065299-88065299 on Build GRCh38
rs374053434 Chr.16: 88065155-88065155 on Build GRCh38
rs765715551 Chr.16: 88065282-88065282 on Build GRCh38
rs1038466894 Chr.16: 88065154-88065154 on Build GRCh38
rs769835310 Chr.16: 88065237-88065237 on Build GRCh38
rs753275873 Chr.16: 88065293-88065293 on Build GRCh38
rs929909968 Chr.16: 88065189-88065189 on Build GRCh38
rs543410643 Chr.16: 88065275-88065275 on Build GRCh38
rs781381240 Chr.16: 88065227-88065227 on Build GRCh38
rs930331640 Chr.16: 88065213-88065213 on Build GRCh38
rs868242731 Chr.16: 88065130-88065130 on Build GRCh38
rs561626806 Chr.16: 88065279-88065279 on Build GRCh38
rs759958359 Chr.16: 88065280-88065280 on Build GRCh38
rs775296755 Chr.16: 88065241-88065241 on Build GRCh38
rs374607381 Chr.16: 88065231-88065231 on Build GRCh38
rs564390753 Chr.16: 88065162-88065162 on Build GRCh38
rs758792003 Chr.16: 88065296-88065296 on Build GRCh38
rs529025986 Chr.16: 88065319-88065319 on Build GRCh38
rs980924295 Chr.16: 88065123-88065123 on Build GRCh38
rs773953331 Chr.16: 88065266-88065266 on Build GRCh38
rs971310949 Chr.16: 88065114-88065114 on Build GRCh38
rs751808800 Chr.16: 88065305-88065305 on Build GRCh38
rs757357170 Chr.16: 88065308-88065308 on Build GRCh38
rs961143335 Chr.16: 88065141-88065141 on Build GRCh38
rs1047019529 Chr.16: 88065221-88065221 on Build GRCh38
rs375667457 Chr.16: 88065309-88065309 on Build GRCh38
rs761497758 Chr.16: 88065267-88065267 on Build GRCh38

View Full Product Details