Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139414081-139414340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC18
Entrez Gene ID
202052
Gene Name
DnaJ heat shock protein family (Hsp40) member C18
Gene Aliases
-
UniGene
Hs.483537
Gene Chromosome Location
Chr.5: 139410203-139439525 on Build GRCh38
Transcripts
NM_152686.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6647288 c.976G>A p.G326R Chr.5: 139414249-139414249 on Build GRCh38
482024 c.974C>T p.A325V Chr.5: 139414251-139414251 on Build GRCh38
1696229 c.1027G>A p.E343K Chr.5: 139414198-139414198 on Build GRCh38
1163380 c.1012G>A p.E338K Chr.5: 139414213-139414213 on Build GRCh38
6647286 c.1009G>A p.A337T Chr.5: 139414216-139414216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372536967 Chr.5: 139414130-139414130 on Build GRCh38
rs926372574 Chr.5: 139414289-139414289 on Build GRCh38
rs748334533 Chr.5: 139414184-139414184 on Build GRCh38
rs779145056 Chr.5: 139414160-139414161 on Build GRCh38
rs201783854 Chr.5: 139414208-139414208 on Build GRCh38
rs774034517 Chr.5: 139414174-139414174 on Build GRCh38
rs374143968 Chr.5: 139414238-139414238 on Build GRCh38
rs369684305 Chr.5: 139414162-139414162 on Build GRCh38
rs764365674 Chr.5: 139414230-139414230 on Build GRCh38
rs968262995 Chr.5: 139414125-139414125 on Build GRCh38
rs752628347 Chr.5: 139414120-139414120 on Build GRCh38
rs746139296 Chr.5: 139414247-139414247 on Build GRCh38
rs767320675 Chr.5: 139414189-139414189 on Build GRCh38
rs780573497 Chr.5: 139414279-139414279 on Build GRCh38
rs377218182 Chr.5: 139414161-139414161 on Build GRCh38
rs779332643 Chr.5: 139414149-139414149 on Build GRCh38
rs14764 Chr.5: 139414193-139414193 on Build GRCh38
rs762583023 Chr.5: 139414163-139414163 on Build GRCh38
rs531443491 Chr.5: 139414144-139414144 on Build GRCh38
rs868460495 Chr.5: 139414213-139414213 on Build GRCh38
rs759306645 Chr.5: 139414180-139414180 on Build GRCh38
rs752639100 Chr.5: 139414191-139414191 on Build GRCh38
rs184813104 Chr.5: 139414159-139414159 on Build GRCh38
rs746227609 Chr.5: 139414151-139414151 on Build GRCh38
rs764176827 Chr.5: 139414223-139414223 on Build GRCh38
rs375386059 Chr.5: 139414296-139414296 on Build GRCh38
rs775871165 Chr.5: 139414152-139414152 on Build GRCh38
rs1056820255 Chr.5: 139414231-139414231 on Build GRCh38
rs142551383 Chr.5: 139414154-139414154 on Build GRCh38
rs146040784 Chr.5: 139414155-139414155 on Build GRCh38
rs772498479 Chr.5: 139414113-139414113 on Build GRCh38
rs1041568540 Chr.5: 139414286-139414286 on Build GRCh38
rs937723047 Chr.5: 139414291-139414291 on Build GRCh38
rs758798306 Chr.5: 139414274-139414274 on Build GRCh38
rs756167364 Chr.5: 139414128-139414128 on Build GRCh38
rs757475729 Chr.5: 139414236-139414236 on Build GRCh38
rs1056064257 Chr.5: 139414299-139414299 on Build GRCh38
rs772416811 Chr.5: 139414189-139414190 on Build GRCh38
rs541497531 Chr.5: 139414250-139414250 on Build GRCh38
rs1020798249 Chr.5: 139414111-139414111 on Build GRCh38
rs769033816 Chr.5: 139414307-139414307 on Build GRCh38
rs749459331 Chr.5: 139414131-139414131 on Build GRCh38
rs766015799 Chr.5: 139414165-139414165 on Build GRCh38

View Full Product Details