Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169780585-169780856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYNN
Entrez Gene ID
55892
Gene Name
myoneurin
Gene Aliases
OSZF, SBBIZ1, ZBTB31, ZNF902
UniGene
Hs.507025
Gene Chromosome Location
Chr.3: 169772247-169789716 on Build GRCh38
Transcripts
NM_001185118.1 NM_001185119.1 NM_018657.4 NR_033702.1 NR_033703.1 XM_005247621.4 XM_005247622.3 XM_005247624.3 XM_011512987.1 XM_017006864.1 XM_017006865.1 XM_017006866.1 XM_017006867.1 XM_017006868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6406576 c.1196C>A p.S399Y Chr.3: 169780725-169780725 on Build GRCh38
6883813 c.1096G>A p.G366R Chr.3: 169780625-169780625 on Build GRCh38
7266133 c.1155A>C p.K385N Chr.3: 169780684-169780684 on Build GRCh38
4149656 c.1122C>G p.V374V Chr.3: 169780651-169780651 on Build GRCh38
4795295 c.1116G>A p.Q372Q Chr.3: 169780645-169780645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763099051 Chr.3: 169780742-169780742 on Build GRCh38
rs767326680 Chr.3: 169780761-169780761 on Build GRCh38
rs767228827 Chr.3: 169780620-169780620 on Build GRCh38
rs139138672 Chr.3: 169780645-169780645 on Build GRCh38
rs759319161 Chr.3: 169780612-169780612 on Build GRCh38
rs769011008 Chr.3: 169780693-169780693 on Build GRCh38
rs760709880 Chr.3: 169780778-169780778 on Build GRCh38
rs898253487 Chr.3: 169780777-169780777 on Build GRCh38
rs371919583 Chr.3: 169780796-169780796 on Build GRCh38
rs748160662 Chr.3: 169780708-169780708 on Build GRCh38
rs778697000 Chr.3: 169780657-169780657 on Build GRCh38
rs143898480 Chr.3: 169780712-169780712 on Build GRCh38
rs367946936 Chr.3: 169780703-169780703 on Build GRCh38
rs772569408 Chr.3: 169780661-169780661 on Build GRCh38
rs570023697 Chr.3: 169780762-169780762 on Build GRCh38
rs750575228 Chr.3: 169780659-169780659 on Build GRCh38
rs777040598 Chr.3: 169780775-169780782 on Build GRCh38
rs567143440 Chr.3: 169780654-169780654 on Build GRCh38
rs776819666 Chr.3: 169780700-169780700 on Build GRCh38
rs747581519 Chr.3: 169780689-169780689 on Build GRCh38
rs994357488 Chr.3: 169780794-169780794 on Build GRCh38
rs866125549 Chr.3: 169780711-169780711 on Build GRCh38
rs773422274 Chr.3: 169780737-169780737 on Build GRCh38
rs755960491 Chr.3: 169780641-169780641 on Build GRCh38
rs763919378 Chr.3: 169780781-169780781 on Build GRCh38
rs780231347 Chr.3: 169780668-169780668 on Build GRCh38
rs752690208 Chr.3: 169780639-169780639 on Build GRCh38
rs1047229535 Chr.3: 169780720-169780720 on Build GRCh38
rs771963538 Chr.3: 169780710-169780711 on Build GRCh38
rs753258214 Chr.3: 169780649-169780649 on Build GRCh38

View Full Product Details