Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157678805-157679032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
UniGene
Hs.292449
Gene Chromosome Location
Chr.1: 157676481-157700985 on Build GRCh38
Transcripts
NM_001320333.1 NM_052939.3 NR_135214.1 NR_135215.1 NR_135216.1 NR_135217.1 XM_006711145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4534409 c.2085G>A p.L695L Chr.1: 157678830-157678830 on Build GRCh38
6340718 c.2106C>A p.D702E Chr.1: 157678809-157678809 on Build GRCh38
7368034 c.2047C>A p.Q683K Chr.1: 157678953-157678953 on Build GRCh38
6307241 c.2073C>A p.L691L Chr.1: 157678842-157678842 on Build GRCh38
4684231 c.2051A>G p.E684G Chr.1: 157678949-157678949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761676311 Chr.1: 157678995-157678995 on Build GRCh38
rs756650560 Chr.1: 157678944-157678944 on Build GRCh38
rs770665069 Chr.1: 157678859-157678859 on Build GRCh38
rs199820507 Chr.1: 157678880-157678880 on Build GRCh38
rs745574624 Chr.1: 157678971-157678971 on Build GRCh38
rs370979735 Chr.1: 157678994-157678994 on Build GRCh38
rs775182398 Chr.1: 157678983-157678983 on Build GRCh38
rs574640324 Chr.1: 157678879-157678880 on Build GRCh38
rs778117882 Chr.1: 157678945-157678945 on Build GRCh38
rs770267838 Chr.1: 157678837-157678837 on Build GRCh38
rs1039984770 Chr.1: 157678923-157678923 on Build GRCh38
rs776539364 Chr.1: 157678915-157678915 on Build GRCh38
rs748261388 Chr.1: 157678825-157678825 on Build GRCh38
rs1052580159 Chr.1: 157678872-157678872 on Build GRCh38
rs944308291 Chr.1: 157678922-157678922 on Build GRCh38
rs748593499 Chr.1: 157678824-157678824 on Build GRCh38
rs764886534 Chr.1: 157678999-157678999 on Build GRCh38
rs749582286 Chr.1: 157678858-157678858 on Build GRCh38
rs146932812 Chr.1: 157678854-157678854 on Build GRCh38
rs142600487 Chr.1: 157678840-157678840 on Build GRCh38
rs944249559 Chr.1: 157678907-157678907 on Build GRCh38
rs749174487 Chr.1: 157678863-157678864 on Build GRCh38
rs761470120 Chr.1: 157678917-157678917 on Build GRCh38
rs768739629 Chr.1: 157678988-157678988 on Build GRCh38
rs759117113 Chr.1: 157678863-157678863 on Build GRCh38
rs755068260 Chr.1: 157678932-157678932 on Build GRCh38
rs199704975 Chr.1: 157678931-157678931 on Build GRCh38
rs912794034 Chr.1: 157678912-157678912 on Build GRCh38
rs749683216 Chr.1: 157678956-157678956 on Build GRCh38
rs935106798 Chr.1: 157678849-157678849 on Build GRCh38
rs771640741 Chr.1: 157678878-157678878 on Build GRCh38
rs766657529 Chr.1: 157678919-157678919 on Build GRCh38
rs549241710 Chr.1: 157678848-157678848 on Build GRCh38
rs778631209 Chr.1: 157678970-157678970 on Build GRCh38
rs961449103 Chr.1: 157678836-157678836 on Build GRCh38
rs746497533 Chr.1: 157678985-157678985 on Build GRCh38
rs764094660 Chr.1: 157678886-157678886 on Build GRCh38
rs6683713 Chr.1: 157678936-157678936 on Build GRCh38
rs771846968 Chr.1: 157678972-157678972 on Build GRCh38
rs549842882 Chr.1: 157678870-157678870 on Build GRCh38
rs757521851 Chr.1: 157678962-157678962 on Build GRCh38
rs900121290 Chr.1: 157678920-157678920 on Build GRCh38

View Full Product Details