Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15354830-15355103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP8
Entrez Gene ID
10270
Gene Name
A-kinase anchoring protein 8
Gene Aliases
AKAP 95, AKAP-8, AKAP-95, AKAP95
UniGene
Hs.594496
Gene Chromosome Location
Chr.19: 15353521-15379815 on Build GRCh38
Transcripts
NM_005858.3 XM_011527624.2 XM_017026141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
460040 c.1997G>C p.R666T Chr.19: 15354997-15354997 on Build GRCh38
709703 c.2050G>C p.E684Q Chr.19: 15354944-15354944 on Build GRCh38
6597799 c.1920C>A p.P640P Chr.19: 15355074-15355074 on Build GRCh38
709702 c.1942G>A p.E648K Chr.19: 15355052-15355052 on Build GRCh38
5570241 c.1898C>T p.T633M Chr.19: 15355096-15355096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985785885 Chr.19: 15354983-15354983 on Build GRCh38
rs921463507 Chr.19: 15355008-15355008 on Build GRCh38
rs780636707 Chr.19: 15354972-15354972 on Build GRCh38
rs564117969 Chr.19: 15354872-15354872 on Build GRCh38
rs767131288 Chr.19: 15354896-15354896 on Build GRCh38
rs777457097 Chr.19: 15355079-15355079 on Build GRCh38
rs1005590984 Chr.19: 15354858-15354858 on Build GRCh38
rs761457640 Chr.19: 15354975-15354983 on Build GRCh38
rs376489439 Chr.19: 15354887-15354887 on Build GRCh38
rs576732307 Chr.19: 15354871-15354871 on Build GRCh38
rs149974098 Chr.19: 15354975-15354975 on Build GRCh38
rs754744442 Chr.19: 15354924-15354924 on Build GRCh38
rs764405941 Chr.19: 15355022-15355022 on Build GRCh38
rs753525803 Chr.19: 15354914-15354914 on Build GRCh38
rs768511127 Chr.19: 15354946-15354946 on Build GRCh38
rs774129123 Chr.19: 15354888-15354888 on Build GRCh38
rs763327144 Chr.19: 15355077-15355077 on Build GRCh38
rs758673111 Chr.19: 15354931-15354931 on Build GRCh38
rs947540357 Chr.19: 15355013-15355013 on Build GRCh38
rs145842466 Chr.19: 15354929-15354929 on Build GRCh38
rs758440981 Chr.19: 15354865-15354865 on Build GRCh38
rs757662753 Chr.19: 15354939-15354939 on Build GRCh38
rs767226233 Chr.19: 15354995-15354996 on Build GRCh38
rs749222352 Chr.19: 15354932-15354932 on Build GRCh38
rs201831883 Chr.19: 15354948-15354948 on Build GRCh38
rs752498693 Chr.19: 15355010-15355010 on Build GRCh38
rs369631726 Chr.19: 15354997-15354997 on Build GRCh38
rs560315178 Chr.19: 15354973-15354973 on Build GRCh38
rs772837580 Chr.19: 15354897-15354897 on Build GRCh38
rs774137622 Chr.19: 15354897-15354897 on Build GRCh38
rs776598548 Chr.19: 15354995-15354995 on Build GRCh38
rs373178064 Chr.19: 15354999-15354999 on Build GRCh38
rs866119603 Chr.19: 15355039-15355039 on Build GRCh38
rs139907510 Chr.19: 15354974-15354974 on Build GRCh38
rs746277361 Chr.19: 15354886-15354886 on Build GRCh38
rs756743031 Chr.19: 15355038-15355038 on Build GRCh38
rs201090278 Chr.19: 15355032-15355032 on Build GRCh38
rs538149553 Chr.19: 15355080-15355080 on Build GRCh38
rs749675913 Chr.19: 15355044-15355044 on Build GRCh38
rs760473339 Chr.19: 15354898-15354898 on Build GRCh38
rs780508322 Chr.19: 15355041-15355041 on Build GRCh38
rs377076421 Chr.19: 15354942-15354942 on Build GRCh38
rs765056046 Chr.19: 15354925-15354925 on Build GRCh38
rs763092973 Chr.19: 15355016-15355016 on Build GRCh38
rs747113664 Chr.19: 15354933-15354933 on Build GRCh38
rs779601230 Chr.19: 15354870-15354870 on Build GRCh38
rs377325571 Chr.19: 15354913-15354913 on Build GRCh38
rs145418809 Chr.19: 15355034-15355034 on Build GRCh38
rs560007341 Chr.19: 15354904-15354904 on Build GRCh38
rs760265810 Chr.19: 15354982-15354982 on Build GRCh38
rs781734939 Chr.19: 15354940-15354940 on Build GRCh38
rs770394900 Chr.19: 15354984-15354984 on Build GRCh38
rs771586432 Chr.19: 15355051-15355051 on Build GRCh38
rs200376540 Chr.19: 15355035-15355035 on Build GRCh38
rs761601749 Chr.19: 15354890-15354890 on Build GRCh38
rs569129129 Chr.19: 15355037-15355037 on Build GRCh38

View Full Product Details