Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43623864-43624101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP2
Entrez Gene ID
54676
Gene Name
GTP binding protein 2
Gene Aliases
-
UniGene
Hs.485449
Gene Chromosome Location
Chr.6: 43620481-43629246 on Build GRCh38
Transcripts
NM_001286216.1 NM_019096.4 XM_017010976.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
335244 c.1207G>T p.E403* Chr.6: 43623962-43623962 on Build GRCh38
1685603 c.1139_1142delAGAG p.E380fs*9 Chr.6: 43624027-43624030 on Build GRCh38
4643096 c.1129G>A p.V377M Chr.6: 43624040-43624040 on Build GRCh38
53521 c.1199A>T p.E400V Chr.6: 43623970-43623970 on Build GRCh38
6568928 c.1220_1222delAGC p.Q407delQ Chr.6: 43623947-43623949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750685958 Chr.6: 43623891-43623891 on Build GRCh38
rs1023819078 Chr.6: 43624034-43624034 on Build GRCh38
rs760866397 Chr.6: 43623893-43623893 on Build GRCh38
rs745811372 Chr.6: 43623954-43623955 on Build GRCh38
rs373891658 Chr.6: 43624067-43624067 on Build GRCh38
rs758165889 Chr.6: 43623943-43623943 on Build GRCh38
rs544214009 Chr.6: 43623918-43623918 on Build GRCh38
rs773554942 Chr.6: 43624079-43624079 on Build GRCh38
rs902925302 Chr.6: 43623896-43623896 on Build GRCh38
rs777686283 Chr.6: 43623987-43623987 on Build GRCh38
rs993963623 Chr.6: 43624023-43624023 on Build GRCh38
rs199987351 Chr.6: 43624071-43624071 on Build GRCh38
rs774685089 Chr.6: 43624059-43624059 on Build GRCh38
rs769233241 Chr.6: 43624044-43624044 on Build GRCh38
rs766504920 Chr.6: 43623897-43623897 on Build GRCh38
rs144104693 Chr.6: 43623990-43623990 on Build GRCh38
rs753911137 Chr.6: 43623904-43623904 on Build GRCh38
rs2232215 Chr.6: 43623885-43623885 on Build GRCh38
rs370688697 Chr.6: 43623908-43623908 on Build GRCh38
rs778667299 Chr.6: 43623913-43623913 on Build GRCh38
rs1037959943 Chr.6: 43623963-43623963 on Build GRCh38
rs745312503 Chr.6: 43624019-43624019 on Build GRCh38
rs780851275 Chr.6: 43624005-43624005 on Build GRCh38
rs905761185 Chr.6: 43623923-43623923 on Build GRCh38
rs147323043 Chr.6: 43623991-43623991 on Build GRCh38

View Full Product Details