Gene Transcripts

Gene Symbol
Ap1g1
Entrez Gene ID
11765
Gene Name
adaptor protein complex AP-1, gamma 1 subunit
Gene Aliases
AA409002, AU041323, AW551707, Adtg, D8Ertd374e
Chromosome Location
Chr. 8: 109778583 - 109864210 on Build GRCm38
UniGene ID
Mm.37210
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301211.1 NP_001288140.1 12 1527
NM_009677.6 NP_033807.2 13 1536
GenBank mRNA AK034198.1 - 10 765
AK036387.1 - 13 1479
AK145821.1 - 12 1458
BC006691.1 - 12 1203
BC052703.1 - 13 1409 5698075
BC054535.1 - 13 1409 5698075
X54424.1 - 12 1449

View Full Product Details