Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009827.2 | NP_033957.1 | 3 | 860 | |
GenBank mRNA | AK004730.2 | - | 3 | 718 | |
AK143895.1 | - | 3 | 708 | ||
AK146738.1 | - | 3 | 861 | ||
AK147052.1 | - | 3 | 861 | ||
AK164301.1 | - | 452 | |||
BC020534.1 | - | 3 | 680 | 4236240 |