Gene Transcripts

Gene Symbol
Pcdha5
Entrez Gene ID
12941
Gene Name
protocadherin alpha 5
Gene Aliases
Cnr6, Crnr6
Chromosome Location
Chr. 18: 36960440 - 37187657 on Build GRCm38
UniGene ID
Mm.308500
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009959.1 NP_034089.1 1 725
GenBank mRNA AB008181.1 - 1 618
AY013763.1 - 1 725

View Full Product Details