Gene Transcripts
- Gene Symbol
- Jak3
- Entrez Gene ID
- 16453
- Gene Name
- Janus kinase 3
- Gene Aliases
- fae, wil
- Chromosome Location
- Chr. 8: 71676383 - 71690577 on Build GRCm38
- UniGene ID
- Mm.249645
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190830.1 |
NP_001177759.1
|
12, 13 |
1890 |
|
NM_010589.6 |
NP_034719.2
|
12, 13 |
1890 |
|
GenBank mRNA |
AK038268.1 |
- |
11, 12 |
1701 |
|
AK088365.1 |
- |
12, 13 |
1739 |
|
AK142178.1 |
- |
12, 13 |
1728 |
|
AK164520.1 |
- |
12, 13 |
1730 |
|
BC105577.1 |
- |
11, 12 |
1693 |
3968386 |
BC131646.1 |
- |
11, 12 |
1701 |
40110146 |
BC131647.1 |
- |
11, 12 |
1701 |
40110148 |
L32955.1 |
- |
11, 12 |
1689 |
|
L33768.1 |
- |
9, 10 |
2331 |
|
L40172.1 |
- |
12, 13 |
1751 |
|
View Full Product Details