Gene Transcripts
- Gene Symbol
- Nek1
- Entrez Gene ID
- 18004
- Gene Name
- NIMA (never in mitosis gene a)-related expressed kinase 1
- Gene Aliases
- D8Ertd790e, kat
- Chromosome Location
- Chr. 8: 60993193 - 61131349 on Build GRCm38
- UniGene ID
- Mm.486881
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001293637.1 |
NP_001280566.1
|
23, 24 |
2739 |
|
NM_001293638.1 |
NP_001280567.1
|
22, 23 |
2607 |
|
NM_001293639.1 |
NP_001280568.1
|
20, 21 |
2448 |
|
NM_175089.4 |
NP_780298.2
|
21, 22 |
2523 |
|
GenBank mRNA |
AK009484.1 |
- |
15, 16 |
1494 |
|
AK010196.1 |
- |
15, 16 |
1494 |
|
AK077047.1 |
- |
22, 23 |
2196 |
|
AK173292.1 |
- |
22, 23 |
2605 |
|
AY850065.1 |
- |
21, 22 |
2496 |
|
BC070443.1 |
- |
21, 22 |
2466 |
|
BC157939.1 |
- |
20, 21 |
2376 |
9088119 |
BC158009.1 |
- |
23, 24 |
2667 |
9007317 |
BC172091.1 |
- |
22, 23 |
2535 |
9054377 |
View Full Product Details