Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011465.4 | NP_035595.2 | 36 | 5332 | |
GenBank mRNA | AF093576.1 | - | 36 | 5240 | |
AK161449.1 | - | 7 | 973 | ||
BC150747.1 | - | 36 | 5197 | 9087658 | |
M10276.1 | - | 4 | 469 | ||
U87455.1 | - | 36 | 5312 |