Gene Transcripts

Gene Symbol
Dtx4
Entrez Gene ID
207521
Gene Name
deltex 4 homolog (Drosophila)
Gene Aliases
AI449438, BC044798, D930042H13, RNF155
Chromosome Location
Chr. 19: 12466329 - 12502201 on Build GRCm38
UniGene ID
Mm.247695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172442.3 NP_766030.3 7 2218
GenBank mRNA AK086624.1 - 6 787
BC044798.1 - 2 101
BC058647.1 - 7 1676 6405881
BC069975.1 - 6 758

View Full Product Details