Gene Transcripts

Gene Symbol
Ubxn10
Entrez Gene ID
212190
Gene Name
UBX domain protein 10
Gene Aliases
5730509E04Rik, A830047D02, Ubxd3
Chromosome Location
Chr. 4: 138709837 - 138738438 on Build GRCm38
UniGene ID
Mm.292226
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285928.1 NP_001272857.1 4 574
NM_001285929.1 NP_001272858.1 3 463
NM_001285930.1 NP_001272859.1 3 327
NM_178671.5 NP_848786.1 2 196
GenBank mRNA AK045964.1 - 2 154
AK046949.1 - 4 575
AK077656.1 - 2 198
AK080662.1 - 2 162
AK133185.1 - 2 182
AK161245.1 - 2 183
AK161296.1 - 2 159

View Full Product Details