Gene Transcripts

Gene Symbol
Astl
Entrez Gene ID
215095
Gene Name
astacin-like metalloendopeptidase (M12 family)
Gene Aliases
7420700F21, C87576, Sas1b
Chromosome Location
Chr. 2: 127338639 - 127357655 on Build GRCm38
UniGene ID
Mm.31313
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291003.1 NP_001277932.1 9 980
NM_172539.3 NP_766127.1 9 1111
GenBank mRNA AK033037.1 - 9 1111
BC064729.1 - 9 972 30476764
FJ187790.1 - 9 919
FJ187791.1 - 8 856
FJ187792.1 - 8 754
FJ187793.1 - 9 817
FJ187794.1 - 7 790
FJ187795.1 - 8 853

View Full Product Details