Gene Transcripts

Gene Symbol
Cep76
Entrez Gene ID
225659
Gene Name
centrosomal protein 76
Gene Aliases
6230425F05Rik, 9630013E05
Chromosome Location
Chr. 18: 67617397 - 67641336 on Build GRCm38
UniGene ID
Mm.111928
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081073.1 NP_001074542.1 10 1586
GenBank mRNA AK035879.1 - 9 1366
BC119216.1 - 10 1467 8733965
BC119218.1 - 10 1467 8733967

View Full Product Details