Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173765.3 | NP_776126.1 | 8 | 1440 | |
GenBank mRNA | AB095954.1 | - | 8 | 1457 | |
AK038779.1 | - | 9 | 1450 | ||
AK136849.1 | - | 8 | 1438 | ||
AK148405.1 | - | 8 | 1439 | ||
AK194577.1 | - | 2 | 57 | ||
BC058102.1 | - | 8 | 1425 | ||
BC094507.1 | - | 6 | 1673 | ||
BC128330.1 | - | 7 | 1312 | 40110405 |