Gene Transcripts

Gene Symbol
Olfr679
Entrez Gene ID
259046
Gene Name
olfactory receptor 679
Gene Aliases
MOR40-2
Chromosome Location
Chr. 7: 105085718 - 105086665 on Build GRCm38
UniGene ID
Mm.377805
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147044.1 NP_667255.1 1 862
GenBank mRNA BC127993.1 - 1 866

View Full Product Details