Gene Transcripts

Gene Symbol
Gpa33
Entrez Gene ID
59290
Gene Name
glycoprotein A33 (transmembrane)
Gene Aliases
2010310L10Rik, 2210401D16Rik, BB116197
Chromosome Location
Chr. 1: 166130460 - 166166510 on Build GRCm38
UniGene ID
Mm.46357
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021610.1 NP_067623.1 4 621
GenBank mRNA AF247659.1 - 4 621
BC008528.1 - 4 614 3593917

View Full Product Details