Gene Transcripts

Gene Symbol
Zdhhc12
Entrez Gene ID
66220
Gene Name
zinc finger, DHHC domain containing 12
Gene Aliases
1190004A01Rik
Chromosome Location
Chr. 2: 30090944 - 30093635 on Build GRCm38
UniGene ID
Mm.22538
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037762.1 NP_001032851.1 5 692
NM_025428.2 NP_079704.2 5 734
GenBank mRNA AK004475.1 - 5 635
AK013074.1 - 5 722
AK087676.1 - 5 706
BC021432.1 - 5 674 5030263

View Full Product Details