Gene Transcripts
- Gene Symbol
- Sppl2a
- Entrez Gene ID
- 66552
- Gene Name
- signal peptide peptidase like 2A
- Gene Aliases
- 2010106G01Rik, C130089K23Rik
- Chromosome Location
- Chr. 2: 126890391 - 126933235 on Build GRCm38
- UniGene ID
- Mm.269928
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB041547.1 |
- |
15 |
1736 |
|
View Full Product Details