Gene Transcripts

Gene Symbol
Ppp1r42
Entrez Gene ID
69312
Gene Name
protein phosphatase 1, regulatory subunit 42
Gene Aliases
1700011J18Rik, 4930418G15Rik, Lrrc67, TLRR
Chromosome Location
Chr. 1: 9968622 - 10013541 on Build GRCm38
UniGene ID
Mm.386795
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145692.2 NP_663730.1 4 468
GenBank mRNA AK005871.1 - 4 463
AK015164.1 - 4 463
AK076637.1 - 4 469
BC022722.1 - 4 452 4503172

View Full Product Details