Gene Transcripts

Gene Symbol
Epn3
Entrez Gene ID
71889
Gene Name
epsin 3
Gene Aliases
2310022G12Rik
Chromosome Location
Chr. 11: 94489599 - 94499974 on Build GRCm38
UniGene ID
Mm.248620
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027984.3 NP_082260.1 9 1938
GenBank mRNA AK009469.1 - 8 1318
AK158031.1 - 8 1816
BC016454.1 - 9 1887 4217221

View Full Product Details