Gene Transcripts

Gene Symbol
Morn3
Entrez Gene ID
74890
Gene Name
MORN repeat containing 3
Gene Aliases
4930438O03Rik
Chromosome Location
Chr. 5: 123033392 - 123047036 on Build GRCm38
UniGene ID
Mm.141383
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029112.1 NP_083388.1 3 381
GenBank mRNA AK015339.1 - 3 534
AK077141.1 - 3 578
BC019987.1 - 3 381 4458628

View Full Product Details