Gene Transcripts

Gene Symbol
Slc10a7
Entrez Gene ID
76775
Gene Name
solute carrier family 10 (sodium/bile acid cotransporter family), member 7
Gene Aliases
2410193C02Rik, P7
Chromosome Location
Chr. 8: 78509328 - 78734012 on Build GRCm38
UniGene ID
Mm.334631
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009981.2 NP_001009981.1 4, 5 645
NM_001282108.1 NP_001269037.1 4, 5 645
NM_029736.2 NP_084012.1 4, 5 645
GenBank mRNA AK010834.1 - 4, 5 595
AK040517.1 - 4, 5 524
AK079901.1 - 4, 5 622
AK152752.1 - 4, 5 606
AK152849.1 - 4, 5 606
AK160402.1 - 4, 5 629
AY825926.1 - 4, 5 398
AY825927.1 - 3, 4 315
AY825928.1 - 4, 5 398
BC116666.1 - 4, 5 511 40107077
BC116717.1 - 4, 5 511 40107078

View Full Product Details