Gene Transcripts

Gene Symbol
Lmod2
Entrez Gene ID
93677
Gene Name
leiomodin 2 (cardiac)
Gene Aliases
C-Lmod
Chromosome Location
Chr. 6: 24597771 - 24605414 on Build GRCm38
UniGene ID
Mm.332941
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053098.2 NP_444328.1 2 818
GenBank mRNA AF237628.1 - 1 55
AK147141.1 - 2 819

View Full Product Details