Gene Transcripts

Gene Symbol
Htr5b
Entrez Gene ID
15564
Gene Name
5-hydroxytryptamine (serotonin) receptor 5B
Gene Aliases
5-Ht5b
Chromosome Location
Chr. 1: 121509758 - 121529312 on Build GRCm38
UniGene ID
Mm.4833
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010483.3 NP_034613.2 1 669
GenBank mRNA BC114988.1 - 1 558
BC115825.1 - 1 558 40056204
X69867.1 - 1 704

View Full Product Details