Gene Transcripts

Gene Symbol
Snta1
Entrez Gene ID
20648
Gene Name
syntrophin, acidic 1
Gene Aliases
AW228934, Snt1
Chromosome Location
Chr. 2: 154376314 - 154408084 on Build GRCm38
UniGene ID
Mm.1541
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009228.2 NP_033254.2 2 509
GenBank mRNA AK037183.1 - 2 407
AK052932.1 - 1 2803
AK137379.1 - 2 510
BB625367.1 - 2 502
BC018546.1 - 2 459 3674921
U00677.1 - 2 506

View Full Product Details