Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175486.3 | NP_780695.1 | 3 | 536 | |
GenBank mRNA | AK032358.1 | - | 3 | 537 | |
AK032501.1 | - | 3 | 543 | ||
AK043338.1 | - | 3 | 576 | ||
AK085251.1 | - | 3 | 567 | ||
BC115775.1 | - | 3 | 448 | 40107115 | |
BC116708.1 | - | 3 | 448 | 40107114 |