Gene Transcripts

Gene Symbol
Slc2a13
Entrez Gene ID
239606
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 13
Gene Aliases
6530403A04, A630029G22Rik, AI505012, Gm308
Chromosome Location
Chr. 15: 91267691 - 91573261 on Build GRCm38
UniGene ID
Mm.360596
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033633.3 NP_001028805.2 2 811
GenBank mRNA AJ874111.1 - 1 131
AK147341.1 - 2 812
BC119586.1 - 3 378 40058865
BC119587.1 - 2 320
BC131681.1 - 2 215

View Full Product Details