Gene Transcripts

Gene Symbol
Gdf6
Entrez Gene ID
242316
Gene Name
growth differentiation factor 6
Gene Aliases
BMP-13, BMP13, GDF16
Chromosome Location
Chr. 4: 9844372 - 9862345 on Build GRCm38
UniGene ID
Mm.302555
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013526.1 NP_038554.1 2 951
GenBank mRNA AJ537425.1 - 2 951
BC141339.1 - 2 865 9055951
BC141340.1 - 2 865 9055952

View Full Product Details