Gene Transcripts
- Gene Symbol
- Fam135b
- Entrez Gene ID
- 70363
- Gene Name
- family with sequence similarity 135, member B
- Gene Aliases
- 1700010C24Rik, A830008O07Rik
- Chromosome Location
- Chr. 15: 71438951 - 71728436 on Build GRCm38
- UniGene ID
- Mm.126450
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043571.1 |
- |
9 |
1094 |
|
BC125645.1 |
- |
9 |
968 |
|
BC125647.1 |
- |
9 |
968 |
|
View Full Product Details