Gene Transcripts

Gene Symbol
Ccdc185
Entrez Gene ID
433386
Gene Name
coiled-coil domain containing 185
Gene Aliases
4922505E12Rik
Chromosome Location
Chr. 1: 182747126 - 182749180 on Build GRCm38
UniGene ID
Mm.338103
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033547.2 NP_001028719.1 1 1529
GenBank mRNA AK077097.1 - 1 837
AK132754.1 - 1 1530
BC150917.1 - 1 1500 9087828

View Full Product Details