Gene Transcripts

Gene Symbol
Zfp397
Entrez Gene ID
69256
Gene Name
zinc finger protein 397
Gene Aliases
2810411K16Rik, 6720480F11Rik, AI461664
Chromosome Location
Chr. 18: 23954662 - 23964671 on Build GRCm38
UniGene ID
Mm.218343
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027007.2 NP_081283.2 4 1135
GenBank mRNA AK013086.1 - 4 1098
AK032001.1 - 4 1138
AY338468.1 - 3 959
BC021456.1 - 4 1119

View Full Product Details