Gene Transcripts
- Gene Symbol
- Mthfd2
- Entrez Gene ID
- 17768
- Gene Name
- methylenetetrahydrofolate dehydrogenase (NAD+ dependent), methenyltetrahydrofolate cyclohydrolase
- Gene Aliases
- AW558851, NMDMC
- Chromosome Location
- Chr. 6: 83305704 - 83325947 on Build GRCm38
- UniGene ID
- Mm.443
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008638.2 |
NP_032664.1
|
4, 5 |
627 |
|
GenBank mRNA |
AK076019.1 |
- |
4, 5 |
580 |
|
AK152249.1 |
- |
4, 5 |
619 |
|
AK159680.1 |
- |
4, 5 |
600 |
|
AK165840.1 |
- |
4, 5 |
619 |
|
AK166262.1 |
- |
4, 5 |
611 |
|
AK196703.1 |
- |
1, 2 |
71 |
|
AK202776.1 |
- |
1, 2 |
70 |
|
BC019511.1 |
- |
4, 5 |
614 |
4164253 |
J04627.1 |
- |
4, 5 |
570 |
|
View Full Product Details