Gene Transcripts
- Gene Symbol
- Cox18
- Entrez Gene ID
- 231430
- Gene Name
- cytochrome c oxidase assembly protein 18
- Gene Aliases
- BC038311
- Chromosome Location
- Chr. 5: 90214724 - 90223996 on Build GRCm38
- UniGene ID
- Mm.319697
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033310.3 |
NP_001028482.2
|
2, 3 |
477 |
|
NM_001163456.1 |
NP_001156928.1
|
2, 3 |
477 |
|
GenBank mRNA |
AK156053.1 |
- |
2, 3 |
446 |
|
BC038311.1 |
- |
2, 3 |
247 |
|
BC138972.1 |
- |
2, 3 |
455 |
8861994 |
BC145479.1 |
- |
2, 3 |
455 |
9054042 |
BC145486.1 |
- |
2, 3 |
459 |
|
BC145728.1 |
- |
2, 3 |
455 |
40131031 |
CX225681.1 |
- |
2, 3 |
404 |
|
DN174415.1 |
- |
2, 3 |
452 |
|
DV661727.1 |
- |
1, 2 |
88 |
|
View Full Product Details