Gene Transcripts

Gene Symbol
Rgs22
Entrez Gene ID
626596
Gene Name
regulator of G-protein signalling 22
Gene Aliases
Chromosome Location
Chr. 15: 35960511 - 36140630 on Build GRCm38
UniGene ID
Mm.321798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195748.1 NP_001182677.1 24 3615
GenBank mRNA CJ236325.1 - 3 402

View Full Product Details