Gene Transcripts
- Gene Symbol
- Tmem9
- Entrez Gene ID
- 66241
- Gene Name
- transmembrane protein 9
- Gene Aliases
- 1500015G18Rik, AW545782
- Chromosome Location
- Chr. 1: 136008219 - 136035030 on Build GRCm38
- UniGene ID
- Mm.41773
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001160145.1 |
NP_001153617.1
|
4, 5 |
494 |
|
NM_001160146.1 |
NP_001153618.1
|
4, 5 |
390 |
|
NM_025439.3 |
NP_079715.1
|
4, 5 |
359 |
|
GenBank mRNA |
AK005248.1 |
- |
4, 5 |
346 |
|
AK021225.1 |
- |
4, 5 |
346 |
|
AK134909.1 |
- |
4, 5 |
324 |
|
AK154423.1 |
- |
4, 5 |
346 |
|
AK218177.1 |
- |
2, 3 |
161 |
|
BC016524.1 |
- |
4, 5 |
334 |
3594475 |
BC020291.1 |
- |
3, 4 |
311 |
|
BC048017.1 |
- |
4, 5 |
313 |
|
BQ918002.1 |
- |
4, 5 |
581 |
|
CA460035.1 |
- |
4, 5 |
396 |
|
CX208138.1 |
- |
4, 5 |
390 |
|
View Full Product Details