Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243043.1 | NP_001229972.1 | 18 | 2842 | |
NM_001243044.1 | NP_001229973.1 | 17 | 2624 | ||
NM_007454.3 | NP_031480.2 | 17 | 2624 | ||
GenBank mRNA | AA003413.1 | - | 3 | 260 | |
AK034140.1 | - | 18 | 2845 | ||
AK155892.1 | - | 17 | 2544 | ||
AK159273.1 | - | 17 | 2515 | ||
AK160042.1 | - | 17 | 2515 | ||
AK165747.1 | - | 17 | 2543 | ||
AK170376.1 | - | 17 | 2535 | ||
BC008513.1 | - | 17 | 2437 | 3601435 | |
BC066827.1 | - | 8 | 1168 | ||
Y07919.1 | - | 17 | 2479 |