Gene Transcripts

Gene Symbol
Ankrd33
Entrez Gene ID
208258
Gene Name
ankyrin repeat domain 33
Gene Aliases
A930021G21Rik, Panky, Panky-a
Chromosome Location
Chr. 15: 101115755 - 101120407 on Build GRCm38
UniGene ID
Mm.152952
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144790.1 NP_659039.1 3 368
GenBank mRNA AK044543.1 - 4 553
AK044546.1 - 3 346
BC022726.1 - 3 368 4504539
FJ895380.1 - 4 529

View Full Product Details