Gene Transcripts

Gene Symbol
Nckap5
Entrez Gene ID
210356
Gene Name
NCK-associated protein 5
Gene Aliases
8430408F21, D130011D22Rik, E030049G20Rik, Erih1, Erih2, Gm1548, Nap5
Chromosome Location
Chr. 1: 125913636 - 126830897 on Build GRCm38
UniGene ID
Mm.377234
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081756.1 NP_001075225.1 14 5308
NM_172484.3 NP_766072.2 12 5169
NM_176957.3 NP_795931.2 11 1198
GenBank mRNA AK053243.1 - 9 881
AY957601.1 - 9 4536
AY957602.1 - 10 4728
BC066095.1 - 12 5169 6856184
BC079551.1 - 12 5169 6856184

View Full Product Details