Gene Transcripts

Gene Symbol
Plk5
Entrez Gene ID
216166
Gene Name
polo-like kinase 5
Gene Aliases
6330514A18Rik
Chromosome Location
Chr. 10: 80356459 - 80365489 on Build GRCm38
UniGene ID
Mm.17613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183152.3 NP_898975.2 14 1681
GenBank mRNA BC086684.1 - 9 978
BC099679.1 - 14 1650 6489647
BU505736.1 - 3 290

View Full Product Details