Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_146020.1 | NP_666132.1 | 1 | 207 | |
GenBank mRNA | AK050107.1 | - | 1 | 221 | |
AK050212.1 | - | 1 | 221 | ||
BC025515.1 | - | 1 | 206 | 5309987 | |
BC030735.1 | - | 1 | 207 | 5126172 |