Gene Transcripts

Gene Symbol
Tgfb1i1
Entrez Gene ID
21804
Gene Name
transforming growth factor beta 1 induced transcript 1
Gene Aliases
ARA55, Hic5, TSC-5, hic-5
Chromosome Location
Chr. 7: 128246799 - 128255699 on Build GRCm38
UniGene ID
Mm.3248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289550.1 NP_001276479.1 6 682
NM_001289551.1 NP_001276480.1 6 682
NM_001289552.1 NP_001276481.1 5 539
NM_001289553.1 NP_001276482.1 5 566
GenBank mRNA AK003170.1 - 6 517
AK041233.1 - 6 510
AK085042.1 - 5 408
AK087114.1 - 6 517
AK156423.1 - 6 517
AK158409.1 - 5 391
BC002049.1 - 6 516 3489459
BC056362.1 - 5 1791 6816930
DQ143891.1 - 5 567
DQ143892.1 - 4 353
DQ143893.1 - 4 360
DQ143894.1 - 4 355
DQ143895.1 - 4 771
DQ143896.1 - 4 732
DQ143897.1 - 4 810
DQ143898.1 - 4 759
DQ143899.1 - 3 886
DQ143900.1 - 5 579
L22482.1 - 5 643

View Full Product Details