Gene Transcripts
- Gene Symbol
- Pphln1
- Entrez Gene ID
- 223828
- Gene Name
- periphilin 1
- Gene Aliases
- CR, HSPC206, HSPC232
- Chromosome Location
- Chr. 15: 93398350 - 93491913 on Build GRCm38
- UniGene ID
- Mm.74208
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001083114.1 |
NP_001076583.1
|
3, 4 |
275 |
|
NM_001285863.1 |
NP_001272792.1
|
3, 4 |
404 |
|
NM_001285864.1 |
NP_001272793.1
|
2, 3 |
183 |
|
NM_146062.3 |
NP_666174.2
|
4, 5 |
482 |
|
NM_175363.4 |
NP_780572.1
|
3, 4 |
275 |
|
GenBank mRNA |
AK081606.1 |
- |
3, 4 |
268 |
|
AK150904.1 |
- |
3, 4 |
226 |
|
AK158488.1 |
- |
3, 4 |
405 |
|
AK160138.1 |
- |
3, 4 |
248 |
|
AK168443.1 |
- |
3, 4 |
276 |
|
BC031486.1 |
- |
4, 5 |
435 |
3157250 |
BC150854.2 |
- |
2, 3 |
179 |
9087765 |
BG145871.1 |
- |
4, 5 |
404 |
|
BY006733.1 |
- |
3, 4 |
276 |
|
CF734096.1 |
- |
2, 3 |
163 |
|
View Full Product Details