Gene Transcripts

Gene Symbol
Tmem63b
Entrez Gene ID
224807
Gene Name
transmembrane protein 63b
Gene Aliases
BC026370
Chromosome Location
Chr. 17: 45660172 - 45686254 on Build GRCm38
UniGene ID
Mm.157904
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198167.3 NP_937810.2 7 603
GenBank mRNA AK090200.1 - 7 524
AK159673.1 - 7 608

View Full Product Details