Gene Transcripts
- Gene Symbol
- Kcnt2
- Entrez Gene ID
- 240776
- Gene Name
- potassium channel, subfamily T, member 2
- Gene Aliases
- E330038N15Rik, Slick
- Chromosome Location
- Chr. 1: 140245659 - 140613962 on Build GRCm38
- UniGene ID
- Mm.483845
- Species
- Mus musculus
- Gene Symbol
- 4930590L20Rik
- Entrez Gene ID
- 75910
- Gene Name
- RIKEN cDNA 4930590L20 gene
- Gene Aliases
- Chromosome Location
- Chr. 1: 140570176 - 140656402 on Build GRCm38
- UniGene ID
- Mm.483845
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016382.1 |
- |
4 |
279 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CA752236.1 |
- |
|
189 |
|
Chr. - - [-] |
View Full Product Details