Gene Transcripts
- Gene Symbol
- Mast4
- Entrez Gene ID
- 328329
- Gene Name
- microtubule associated serine/threonine kinase family member 4
- Gene Aliases
- 4930420O11Rik, AI642422, mKIAA0303
- Chromosome Location
- Chr. 13: 102732488 - 103335840 on Build GRCm38
- UniGene ID
- Mm.202606
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175171.3 |
NP_780380.2
|
29 |
5220 |
|
GenBank mRNA |
AK090136.1 |
- |
29 |
5192 |
|
View Full Product Details