Gene Transcripts

Gene Symbol
Dennd4c
Entrez Gene ID
329877
Gene Name
DENN/MADD domain containing 4C
Gene Aliases
1700065A05Rik, AA420392
Chromosome Location
Chr. 4: 86748555 - 86850603 on Build GRCm38
UniGene ID
Mm.273718
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_184088.1 NP_908976.1 13, 14 2326
GenBank mRNA AK155220.1 - 13, 14 2327
BC146602.1 - 12, 13 1951 40099465

View Full Product Details